siRNA Details
| Gene Symbol: IL12A-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108088.1 | 10 | 2273 | |||
| GenBank mRNA | AK097161.1 | - | 11 | 2273 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000497452.5 | - | 10 | 2273 | Chr. 3 - 159913400 - 160207092 [-] on Build GRCh38 | |

IL12A-AS1
Genomic Map