siRNA Details
| Gene Symbol: LOC105377105 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135534.1 | 2 | 3086 | |||
| GenBank mRNA | BC041347.2 | - | 2 | 3069 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000488720.1 | - | 2 | 3087 | Chr. 3 - 58162547 - 58170636 [-] on Build GRCh38 | |

LOC105377105
Genomic Map