siRNA Details
| Gene Symbol: LOC101559451 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103482.1 | 2 | 252 | |||
| GenBank mRNA | BX109211.1 | - | 2 | 252 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000497885.1 | - | 2 | 252 | Chr. 17 - 4704230 - 4705529 [+] on Build GRCh38 | |

LOC101559451
Genomic Map