siRNA Details
Gene Symbol: GATA2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125398.1 | 2 | 482 | |||
GenBank mRNA | BI258722.1 | - | 2 | 228 | ||
BM770141.1 | - | 2 | 218 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: GATA2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125398.1 | 2 | 482 | |||
GenBank mRNA | BI258722.1 | - | 2 | 228 | ||
BM770141.1 | - | 2 | 218 |