siRNA Details
| Gene Symbol: GATA2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125398.1 | 3 | 896 | |||
| GenBank mRNA | BI258722.1 | - | 3 | 641 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GATA2-AS1| Gene Symbol: GATA2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125398.1 | 3 | 896 | |||
| GenBank mRNA | BI258722.1 | - | 3 | 641 | ||