siRNA Details
Gene Symbol: FAM53A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011513404.2 | XP_011511706.1 | 5 | 2111 | ||
GenBank mRNA | AK093972.1 | - | 1 | 2029 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000466692.1 | - | 2 | 430 | Chr. 4 - 1574062 - 1580253 [-] on Build GRCh38 |
