siRNA Details
Gene Symbol: LINC01213 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110167.1 | 3 | 243 | |||
NR_110168.1 | 3 | 196 | ||||
GenBank mRNA | BF676681.1 | - | 3 | 244 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000472890.5 | - | 3 | 196 | Chr. 3 - 150238519 - 150240203 [+] on Build GRCh38 |
