siRNA Details
Gene Symbol: LINC01391 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121649.1 | 3 | 3253 | |||
GenBank mRNA | AK128202.1 | - | 3 | 3253 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000495287.1 | - | 3 | 3253 | Chr. 3 - 138935189 - 138944020 [-] on Build GRCh38 |
