siRNA Details
| Gene Symbol: LOC101928583 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135555.1 | 3 | 1142 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000497988.1 | - | 2 | 1303 | Chr. 3 - 170656562 - 170662123 [+] on Build GRCh38 | |

LOC101928583
Genomic Map