siRNA Details
Gene Symbol: LOC101928583 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135555.1 | 3 | 1993 | |||
GenBank mRNA | BC042089.1 | - | 2 | 2154 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000497988.1 | - | 2 | 2154 | Chr. 3 - 170656562 - 170662123 [+] on Build GRCh38 |
