siRNA Details
| Gene Symbol: LOC105375650 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132986.1 | 2 | 2480 | |||
| NR_132987.1 | 2 | 2430 | ||||
| Gene Symbol: LOC286149 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK093190.1 | - | 2 | 2430 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000501164.2 | - | 2 | 2430 | Chr. 8 - 95066808 - 95072920 [-] on Build GRCh38 | |

LOC105375650
Genomic Map