siRNA Details
Gene Symbol: SMARCA5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104027.1 | 1 | 252 | |||
GenBank mRNA | AK307172.1 | - | 1 | 22 | ||
HY014185.1 | - | 1 | 252 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SMARCA5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104027.1 | 1 | 252 | |||
GenBank mRNA | AK307172.1 | - | 1 | 22 | ||
HY014185.1 | - | 1 | 252 |