siRNA Details
Gene Symbol: RNF139-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108047.1 | 2 | 1271 | |||
GenBank mRNA | CR933665.1 | - | 2 | 1271 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000499418.2 | - | 2 | 1272 | Chr. 8 - 124462485 - 124474564 [-] on Build GRCh38 |
