siRNA Details
Gene Symbol: LOC100996419 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134283.1 | 2 | 1293 | |||
GenBank mRNA | BC040901.1 | - | 2 | 1283 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000499601.2 | - | 2 | 1283 | Chr. 5 - 179859013 - 179861283 [+] on Build GRCh38 |
