siRNA Details
| Gene Symbol: LOC101928075 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110071.1 | 3 | 2666 | |||
| GenBank mRNA | AK093892.1 | - | 3 | 2666 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000500016.1 | - | 3 | 2666 | Chr. 14 - 70608798 - 70641298 [-] on Build GRCh38 | |

LOC101928075
Genomic Map