siRNA Details
| Gene Symbol: LINC01585 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120371.1 | 3 | 715 | |||
| GenBank mRNA | BC038778.1 | - | 3 | 715 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000500496.2 | - | 3 | 715 | Chr. 15 - 90660234 - 90664967 [+] on Build GRCh38 | |

LINC01585
Genomic Map