siRNA Details
Gene Symbol: LINC01585 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120371.1 | 3 | 792 | |||
GenBank mRNA | BC038778.1 | - | 3 | 792 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000500496.2 | - | 3 | 792 | Chr. 15 - 90660234 - 90664967 [+] on Build GRCh38 |
