siRNA Details
| Gene Symbol: NEAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028272.1 | 1 | 1585 | |||
| NR_131012.1 | 1 | 1585 | ||||
| GenBank mRNA | EF177379.1 | - | 1 | 1585 | ||
| GQ859162.1 | - | 1 | 1585 | |||
| U60873.1 | - | 1 | 362 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000499732.2 | - | 2 | 1366 | Chr. 11 - 65422774 - 65424761 [+] on Build GRCh38 | |

NEAT1
Genomic Map