siRNA Details
| Gene Symbol: LOC101929295 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110303.1 | 3 | 1872 | |||
| GenBank mRNA | AK055250.1 | - | 3 | 1872 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000501356.5 | - | 3 | 1872 | Chr. 11 - 95150539 - 95159148 [+] on Build GRCh38 | |

LOC101929295
Genomic Map