siRNA Details
| Gene Symbol: LOC101928100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120430.1 | 4 | 1521 | |||
| GenBank mRNA | AK096314.1 | - | 4 | 1521 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000500682.1 | - | 4 | 1521 | Chr. 12 - 10363769 - 10398506 [+] on Build GRCh38 | |

LOC101928100
Genomic Map