siRNA Details
| Gene Symbol: LOC283387 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC032840.1 | - | 1 | 1114 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000499481.2 | - | 1 | 1107 | Chr. 12 - 57869835 - 57896482 [-] on Build GRCh38 | |

LOC283387
Genomic Map