siRNA Details
Gene Symbol: LOC728339 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC087857.1 | - | 3 | 566 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000501825.2 | - | 3 | 563 | Chr. 4 - 189864262 - 189939837 [-] on Build GRCh38 |
