siRNA Details
Gene Symbol: LOC101927740 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109890.1 | 3 | 1246 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000499583.1 | - | 3 | 1047 | Chr. 5 - 159100682 - 159117478 [+] on Build GRCh38 |
