siRNA Details
| Gene Symbol: LOC101927740 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109890.1 | 3 | 2635 | |||
| GenBank mRNA | AK123543.1 | - | 3 | 2435 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000499583.1 | - | 3 | 2436 | Chr. 5 - 159100682 - 159117478 [+] on Build GRCh38 | |

LOC101927740
Genomic Map