siRNA Details
| Gene Symbol: MIR4300HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120571.1 | 4, 5 | 667 | |||
| GenBank mRNA | BC041900.1 | - | 4, 5 | 667 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000500502.5 | - | 4, 5 | 667 | Chr. 11 - 81879854 - 82403913 [-] on Build GRCh38 | |

MIR4300HG
Genomic Map