siRNA Details
Gene Symbol: LINC00682 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104143.1 | 2 | 1359 | |||
GenBank mRNA | BC025350.1 | - | 2 | 1300 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000499082.2 | - | 2 | 1318 | Chr. 4 - 41879521 - 41882570 [-] on Build GRCh38 |
