siRNA Details
| Gene Symbol: FLJ21408 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK025061.1 | - | 2 | 1060 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000499939.2 | - | 2 | 1060 | Chr. 16 - 27268869 - 27271976 [+] on Build GRCh38 | |

FLJ21408
Genomic Map