siRNA Details
| Gene Symbol: FLJ21408 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037184.1 | 5 | 543 | |||
| GenBank mRNA | BC044845.1 | - | 5 | 543 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000566854.5 | - | 5 | 543 | Chr. 16 - 27268205 - 27290468 [+] on Build GRCh38 | |

FLJ21408
Genomic Map