siRNA Details
Gene Symbol: LOC101928809 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125906.1 | 3 | 1177 | |||
GenBank mRNA | BC043569.1 | - | 3 | 1177 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000501239.2 | - | 3 | 1177 | Chr. 4 - 75980790 - 76005942 [+] on Build GRCh38 |
