siRNA Details
| Gene Symbol: LOC101928809 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125906.1 | 3 | 1489 | |||
| GenBank mRNA | BC043569.1 | - | 3 | 1489 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000501239.2 | - | 3 | 1489 | Chr. 4 - 75980790 - 76005942 [+] on Build GRCh38 | |

LOC101928809
Genomic Map