siRNA Details
| Gene Symbol: LINC01184 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015360.1 | 3 | 450 | |||
| GenBank mRNA | BX648677.1 | - | 3 | 450 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000501702.6 | - | 3 | 450 | Chr. 5 - 128021528 - 128083074 [-] on Build GRCh38 | |

LINC01184
Genomic Map