siRNA Details
| Gene Symbol: LOC100996286 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121627.1 | 3 | 1056 | |||
| NR_121628.1 | 3 | 720 | ||||
| GenBank mRNA | BC040914.1 | - | 3 | 1056 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100996286| Gene Symbol: LOC100996286 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121627.1 | 3 | 1056 | |||
| NR_121628.1 | 3 | 720 | ||||
| GenBank mRNA | BC040914.1 | - | 3 | 1056 | ||