siRNA Details
Gene Symbol: LOC100996286 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121627.1 | 2 | 476 | |||
NR_121628.1 | 2 | 140 | ||||
GenBank mRNA | BC040914.1 | - | 2 | 476 | ||
CB986668.1 | - | 143 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100996286 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121627.1 | 2 | 476 | |||
NR_121628.1 | 2 | 140 | ||||
GenBank mRNA | BC040914.1 | - | 2 | 476 | ||
CB986668.1 | - | 143 |