siRNA Details
| Gene Symbol: LOC286059 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038873.1 | 3 | 632 | |||
| GenBank mRNA | AK024295.1 | - | 3 | 653 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000501897.1 | - | 3 | 632 | Chr. 8 - 23068229 - 23083619 [+] on Build GRCh38 | |

LOC286059
Genomic Map