siRNA Details
| Gene Symbol: RRS1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040434.1 | 3 | 1150 | |||
| GenBank mRNA | AK125958.1 | - | 3 | 1150 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000499642.1 | - | 3 | 1150 | Chr. 8 - 66419589 - 66428977 [-] on Build GRCh38 | |

RRS1-AS1
Genomic Map