siRNA Details
Gene Symbol: LOC101929524 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109946.1 | 3 | 1005 | |||
GenBank mRNA | AK097784.1 | - | 3 | 1005 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000499131.1 | - | 3 | 1005 | Chr. 5 - 16615926 - 16629969 [+] on Build GRCh38 |
