siRNA Details
Gene Symbol: UBR5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125415.1 | 3 | 580 | |||
GenBank mRNA | AK095151.1 | - | 2 | 726 | ||
BQ082210.1 | - | 3 | 541 | |||
DA677213.1 | - | 3 | 580 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: UBR5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125415.1 | 3 | 580 | |||
GenBank mRNA | AK095151.1 | - | 2 | 726 | ||
BQ082210.1 | - | 3 | 541 | |||
DA677213.1 | - | 3 | 580 |