siRNA Details
| Gene Symbol: LOC101928053 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120540.1 | 3 | 610 | |||
| GenBank mRNA | AK056207.1 | - | 3 | 611 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000499765.1 | - | 3 | 610 | Chr. 11 - 10809297 - 10822931 [+] on Build GRCh38 | |

LOC101928053
Genomic Map