siRNA Details
Gene Symbol: LOC728084 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038385.1 | 3 | 2216 | |||
GenBank mRNA | AK055062.1 | - | 3 | 2216 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000500381.2 | - | 3 | 2203 | Chr. 12 - 89010681 - 89019679 [-] on Build GRCh38 |
