siRNA Details
Gene Symbol: LOC105369192 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134236.1 | 5 | 678 | |||
NR_134237.1 | 4 | 602 | ||||
GenBank mRNA | BC027846.1 | - | 5 | 666 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000500009.2 | - | 5 | 666 | Chr. 4 - 88284942 - 88331421 [+] on Build GRCh38 |
