siRNA Details
| Gene Symbol: LOC105369192 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134236.1 | 5 | 490 | |||
| NR_134237.1 | 4 | 414 | ||||
| GenBank mRNA | BC027846.1 | - | 5 | 478 | ||
| DA132869.1 | - | 4 | 414 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000500009.2 | - | 5 | 478 | Chr. 4 - 88284942 - 88331421 [+] on Build GRCh38 | |

LOC105369192
Genomic Map