siRNA Details
| Gene Symbol: LOC283856 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027078.1 | 1 | 1507 | |||
| GenBank mRNA | BC035113.2 | - | 1 | 1507 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000501259.5 | - | 1 | 1507 | Chr. 16 - 56092987 - 56191094 [-] on Build GRCh38 | |

LOC283856
Genomic Map