siRNA Details
Gene Symbol: LOC101928597 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110091.1 | 3 | 1096 | |||
GenBank mRNA | BC033344.1 | - | 3 | 1097 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000443154.3 | - | 3 | 1096 | Chr. 12 - 133032659 - 133037222 [-] on Build GRCh38 |
