siRNA Details
| Gene Symbol: LOC101928597 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110091.1 | 3 | 702 | |||
| GenBank mRNA | BC033344.1 | - | 3 | 702 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000443154.3 | - | 3 | 702 | Chr. 12 - 133032659 - 133037222 [-] on Build GRCh38 | |

LOC101928597
Genomic Map