siRNA Details
| Gene Symbol: LOC101929210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125922.1 | 5 | 244 | |||
| GenBank mRNA | BC014345.1 | - | 5 | 247 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000501965.2 | - | 5 | 244 | Chr. 4 - 94117792 - 94207556 [-] on Build GRCh38 | |

LOC101929210
Genomic Map