siRNA Details
| Gene Symbol: LOC105377348 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131185.1 | 3 | 821 | |||
| NR_131186.1 | 2 | 691 | ||||
| GenBank mRNA | AK093356.1 | - | 2 | 612 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105377348| Gene Symbol: LOC105377348 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131185.1 | 3 | 821 | |||
| NR_131186.1 | 2 | 691 | ||||
| GenBank mRNA | AK093356.1 | - | 2 | 612 | ||