siRNA Details
| Gene Symbol: LOC106699570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_133569.1 | 1 | 1011 | |||
| GenBank mRNA | AL080152.1 | - | 1 | 1011 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000501143.1 | - | 1 | 1011 | Chr. 16 - 66751752 - 66754740 [+] on Build GRCh38 | |

LOC106699570
Genomic Map