siRNA Details
| Gene Symbol: LOC101927282 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110753.1 | 5 | 1624 | |||
| GenBank mRNA | BC035172.1 | - | 5 | 1631 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000500092.6 | - | 5 | 1624 | Chr. 4 - 129724171 - 129771478 [-] on Build GRCh38 | |

LOC101927282
Genomic Map