siRNA Details
| Gene Symbol: BAALC-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109954.1 | 2 | 548 | |||
| GenBank mRNA | AK125733.1 | - | 7 | 1728 | ||
| AK311189.1 | - | 2 | 548 | |||
| DA226722.1 | - | 2 | 549 | |||
| DA333188.1 | - | 2 | 436 | |||
| DT217610.1 | - | 2 | 435 | |||

BAALC-AS1
Genomic Map