siRNA Details
| Gene Symbol: SNHG10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001459.2 | 2 | 1249 | |||
| NR_003138.3 | 1 | 1880 | ||||
| GenBank mRNA | AK097876.1 | - | 1 | 1742 | ||
| BC036576.2 | - | 1 | 1635 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SNHG10| Gene Symbol: SNHG10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001459.2 | 2 | 1249 | |||
| NR_003138.3 | 1 | 1880 | ||||
| GenBank mRNA | AK097876.1 | - | 1 | 1742 | ||
| BC036576.2 | - | 1 | 1635 | |||